Orders LinkContactUsLinkComponent
Cooking thermometer
Cooking thermometer
Cooking thermometer with white sleeve, without PTFE, −10...+60 °C, 1,0 %, 1,0 °C
Katalog # ALLA801.060-GB
Leverandør:   Alla France
Cooking thermometer
Katalog # ALLA801.060-GB
Leverandør:  Alla France
Cooking thermometer
Cooking thermometer
Cooking thermometer with white sleeve, without PTFE, −10...+60 °C, 1,0 %, 1,0 °C
Price
264,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Internal scale glass thermometer: −10 to +60 °C , for baking, milk and wine.
  • White polypropylene alimentary protection sleeve
Specifikationer
  • Beskrivelse:
    Cooking thermometer with white sleeve, without PTFE
  • Nøjagtighed:
    1,0 %
  • Inddeling:
    1,0 °C
  • Område:
    −10...+60 °C