Wipe rolls, Multisoft® Comfort
Cleaning cloth roll, hvid, 380×260 mm, 2 rolls of 570 m (1500 sheets)
Katalog # ZVGA11476
Leverandør: Zellstoff-Vertriebs-GmbH & Co. KG
Gå til:
Product Details & Documents
These cost-efficient cleaning wipes made from 100% recycled cellulose are ideal for cleaning and maintenance tasks in different industries.
- Two ply
- Highly absorbent
Specifikationer
- Pakket:2 rolls of 570 m (1500 sheets)
- Farve:hvid
- Beskrivelse:Cleaning cloth roll
- L×B:380×260 mm