Orders LinkContactUsLinkComponent
Multiple gradient gel caster, Owl™
Multiple gradient gel caster, Owl™
Gel caster, for gel size 250×130×230 mm (W×D×H)
Katalog # 730-0296
Leverandør:   THERMO OWL SCIENTIFIC
Multiple gradient gel caster, Owl™
Katalog # 730-0296
Leverandør:  THERMO OWL SCIENTIFIC
Multiple gradient gel caster, Owl™
Multiple gradient gel caster, Owl™
Gel caster, for gel size 250×130×230 mm (W×D×H)
Price
6.370,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Multiple gradient gel caster for casting one to five gels simultaneously.
Specifikationer
  • Beskrivelse:
    Gel caster, for gel size 250×130×230 mm (W×D×H)
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement clamps
    Replacement clamps
    700-2310
    Log ind for tilgængelighed
    1 * 1 Items - 2.940,00 kr