Orders LinkContactUsLinkComponent
Woulff bottles
Woulff bottles
500 ml
Catalog # WITG5.663.000
Supplier:   witeg Labortechnik
Woulff bottles
Catalog # WITG5.663.000
Supplier:  witeg Labortechnik
Woulff bottles
Price
3.030,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Glass, clear, plastic coated.
  • Clear plastic coated to protect against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Implosion protection
Complete set includes 500 ml bottle with 3 necks (center neck GL 25, side necks GL 18), suction tube with safety valve, analog pressure gauge. Air-suction tube icnludes PTFE needle valve stopcock.
Specifications
  • Kapacitet:
    500 ml
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • WITG5.663.002
    Suction tube with safety valve
    500 ml woulff bottles
    Log in for availability
    1 * 1 Items - kr414.00