Orders LinkContactUsLinkComponent
Snap vials, clean
Snap vials, clean
   Sustainable Options Available
Supplier:   EP SCIENTIFIC
Snap vials, clean
Supplier:  EP SCIENTIFIC
   Sustainable Options Available
Certificates
Snap vials, clean
3 Options Available Below
Product Details & Documents
Clean snap VOA vials feature a snap-off septum guard that protects the septum from exposure to the general environment. The clean snap cap reduces the chance of volatile organic contaminants in the environment from passing through the septum and contaminating the sample during transport and storage. It also keeps the septum clean and free from foreign particles during handling. The clean snap cap easily peels away to provide access to the septum at the time of analysis.
  • Amber borosilicate glass vial is recommended for light sensitive samples
  • Septa permanently bonded to the cap
  • Clean snap closure features a fully integrated, snap-off septum guard that helps protect the septum from chemical and particulate contamination during handling and storage
  • Standard cap dimensions are automation friendly
  • Fully assembled
Specifications for Product Family
Specifications
  • Catalog No:
  • EPSCSS246-0040
  • EPSCSS136-0040
  • EPSCSS236-0040
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Amber
Product Information
  • Beskrivelse
    Kapacitet
    Diameter×H
    Farve
    Kasket inkluderet
    Cap størrelse
    Septum
    Tykkelse
    Availability
    Price
  • Snap vial with VOA and not certificate
    40,00 ml
    28×95 mm
    ravfarvet
    Yes
    24-414
    Bonded
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 Items
    1 * 72 Items - 2.700,00 kr
    2.700,00 kr
    1 * 72 Items
       2.700,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPSCSS246-0040
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning
Clear
Product Information
  • Beskrivelse
    Kapacitet
    Diameter×H
    Farve
    Kasket inkluderet
    Cap størrelse
    Septum
    Tykkelse
    Availability
    Price
  • Snap vial without certificate
    40,00 ml
    28×95 mm
    klar
    Yes
    24-414
    Bonded
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 Items
    1 * 72 Items - 2.440,00 kr
    2.440,00 kr
    1 * 72 Items
       2.440,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPSCSS136-0040
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning
  • Snap vial with VOA and not certificate
    40,00 ml
    28×95 mm
    klar
    Yes
    24-414
    Bonded
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 Items
    1 * 72 Items - 2.700,00 kr
    2.700,00 kr
    1 * 72 Items
       2.700,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPSCSS236-0040
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning