Anti- IgG Antibody (Unconjugated)
BSBTBA1044-1
- Antistof type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Konjugation:Unconjugated
- Krydsadsorption:No
- Størrelse:1 mg
- Pakn:1 mg
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: