Microfiltration assembly, with solvent pick-up adapter, ULTRA-WARE®, KIMBLE®
Catalog # SCERSP953820-1047
Supplier: DWK Life Sciences
Specifications
- Kapacitet:1000 ml
- Beskrivelse:Microfiltration assembly with solvent pick-up adapter and ground joint, 137×430 mm
- Pakn:1
Specifications
About this item
This microfiltration system manufactured from 33 expansion, low extractable borosilicate glass eliminates pouring HPLC solvents into funnels. The solvent pickup adapter draws solvent directly from a reagent bottle through a 47 mm filter and into a filter flask.
- Support base with 40 to 60 micron porosity glass frit
- Integral vacuum connection above the drip tip to prevent contamination of the vacuum line with filtrate droplets
After filtration is complete, the HI-VAC valve on the solvent pickup adapter can be turned off for vacuum degassing.
Existing ULTRA-WARE® and Millipore filtration assemblies are easily converted to in-line filtration/degassing by replacing existing funnels with solvent pickup adapters. They clamp to support bases in the same way that funnels do.
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements
Delivery: System supplied with a solvent pickup adapter, a coarse porosity fritted glass filter base, an inner joint flask and an aluminum clamp.