Snood cap, S896
hvid, One size
Katalog # PRTWS896WHR
Leverandør: Portwest
Gå til:
Product Details & Documents
Designed specifically for catering environments, the snood cap is a perfect choice when working with food. Made from 100% cotton fabric for added comfort and breathability.
- Cooling mesh fabric (polyester mesh) for increased breathability
- Back neck snood offers excellent hair coverage
- Elastic back allows for a comfortable fit
- Washable at up to 60 °C
Weight: 280 g
Specifikationer
- Tøj størrelse:One size
- Farve:hvid
- Cat. No.:PRTWS896WHR
- Vægt:280 g
- Outfit applikationer:Food
- Stof:Cotton/polyester mesh