Capillary columns, DB-1301
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Om denne vare
These columns contain (6%-Cyanopropyl-phenyl) methylpolysiloxane. They are solvent rinsable and have low or mid-polarity ideal for GC.
- Bonded and cross-linked
Max. temperature: −20 to 280/300 °C and −20 to 260/300 °C (for Ø 0,53 mm)
11 tilgængelige muligheder nedenfor
Product Details & Documents
These columns contain (6%-Cyanopropyl-phenyl) methylpolysiloxane. They are solvent rinsable and have low or mid-polarity ideal for GC.
- Bonded and cross-linked
Max. temperature: −20 to 280/300 °C and −20 to 260/300 °C (for Ø 0,53 mm)
Product Family Options
Produktinformation
- Kolonne formFaseFilmtykkelseØ indv.LængdeTilgængelighedPrice
- HEWL122-1332on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL122-1332 - HEWL122-1333on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL122-1333 - HEWL122-1362on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL122-1362 - HEWL122-1363on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL122-1363 - HEWL123-1332on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL123-1332 - HEWL123-1333on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL123-1333 - HEWL123-1363on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL123-1363 - HEWL125-1312on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL125-1312 - HEWL125-1332on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL125-1332 - HEWL125-1333on 7" cageSpecifikationer:
More
Specifications
Cat. No.HEWL125-1333 - HEWL122-1363Eon 5" cageSpecifikationer:
More
Specifications
Cat. No.HEWL122-1363E
This product has been blocked by your organization. Please contact your purchasing department for more information