Orders LinkContactUsLinkComponent
Radial ventilator
Radial ventilator
Supplier:   asecos
Radial ventilator
Supplier:  asecos
Certificates
Radial ventilator
Radial ventilator
2 Options Available Below
Product Details & Documents
Radial ventilator for a safe and approved technical ventilation of safety storage cabinets.
  • Sealed against gas emission
  • PA/GFK console
  • PP housing, PP impeller
  • Airflow: 0 to 250 m³/hour
  • Dimension: 250×225×310 mm
  • Pressure: 240 to 175 Pa
Recommended for mounting outside the working environment due to high noise level.
Product Family Options
Product Information
  • Beskrivelse
    Vægt
    Availability
    Price
  • Radial ventilator with winding shield
    3,5 kg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.640,00 kr
    3.640,00 kr
    1 * 1 Items
       3.640,00 kr
  • Flexible exhaust tube, Ø: 75 mm
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 203,00 kr
    203,00 kr
    1 * 1 m
       203,00 kr