Orders LinkContactUsLinkComponent
behrotest® Clean-up station, CUS 2
behrotest® Clean-up station, CUS 2
Clean-up station, CUS 2
Katalog # BEHRB00217638
Leverandør:   BEHR
behrotest® Clean-up station, CUS 2
Katalog # BEHRB00217638
Leverandør:  BEHR
behrotest® Clean-up station, CUS 2
behrotest® Clean-up station, CUS 2
Clean-up station, CUS 2
Price
4.380,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The complete sample clean up station includes dropping funnel, clean-up column with glass frit, reducing flasks and stand.
The clean up procedure can be done for two samples simultaneously.
Certifications: Corresponds exactly to the provisions of ISO 9377-2.
Specifikationer
  • Beskrivelse:
    Clean-up station
  • Model:
    CUS 2
Accessories
Produktinformation
  • Katalog #
    Model
    Tilgængelighed
    Price
  • Evaporating flask, graduated, 100 ml
    Evaporating flask, graduated, 100 ml
    BEHRB00217642
    KOG 100
    Log ind for tilgængelighed
    1 * 1 Items - 817,00 kr
Produktinformation
  • Katalog #
    Model
    Tilgængelighed
    Price
  • Microseparator, NS 29, 260×50×80 mm
    Microseparator, NS 29, 260×50×80 mm
    BEHRB00217654
    H 13-3/29
    Log ind for tilgængelighed
    1 * 1 Items - 846,00 kr
Produktinformation
  • Katalog #
    Model
    Tilgængelighed
    Price
  • Clean-up column
    Clean-up column
    BEHRB00217643
    CUC 2
    Log ind for tilgængelighed
    1 * 1 Items - 846,00 kr
Produktinformation
  • Katalog #
    Model
    Tilgængelighed
    Price
  • Separating funnel, 60 ml
    Separating funnel, 60 ml
    BEHRB00217641
    TSE
    Log ind for tilgængelighed
    1 * 1 Items - 2.410,00 kr
Produktinformation
  • Katalog #
    Model
    Tilgængelighed
    Price
  • Microseparator, NS 14, 250×70×80 mm
    Microseparator, NS 14, 250×70×80 mm
    BEHRB00217652
    H 13-2/14
    Log ind for tilgængelighed
    1 * 1 Items - 859,00 kr