Orders LinkContactUsLinkComponent
Racks for SPB system tubes
Racks for SPB system tubes
Rack for SPB 50-24 system, 24 (4×6), 50 ml
Katalog # PERCN9308070
Leverandør:   PerkinElmer
Racks for SPB system tubes
Katalog # PERCN9308070
Leverandør:  PerkinElmer
Racks for SPB system tubes
Rack for SPB 50-24 system, 24 (4×6), 50 ml
Price
2.920,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Polycarbonate.
  • With rack lock feature which locks DigiTUBE®s in position within the rack allowing for easy and one-handed closure of screw cap
  • Acid resistant
  • For laboratory usage
Specifikationer
  • Til rør:
    50 ml
  • Ant. huller:
    24 (4×6)
  • Beskrivelse:
    Rack for SPB 50-24 system