600 series Microliter™ syringes
Leverandør: HAMILTON BONADUZ
Certifikater
Om denne vare
The 600 series syringes are a half stroke version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 0,25 μl up to 5 μl. The 600 series is often used in applications that require one handed operation because of the short stroke length and the reinforced plunger.
Specifikationer
- Catalog No:
- HAMI7633-01
- HAMI7632-01
- Væskesti:
- Stainless steel, borosilicate glass
- Stainless steel, borosilicate glass
- Maks. tryk:
- 137,9 bar
- 137,9 bar
- Max. temperature:
- 115 °C
- 115 °C
- Min. temperature:
- 10 °C
- 10 °C