Orders LinkContactUsLinkComponent
EnteroPluri test
EnteroPluri test
Supplier:  LIOFILCHEM
Certificates
EnteroPluri test
EnteroPluri test
17 Options Available Below
Product Details & Documents
EnteroPluri Test is a 12-sector system containing special culture media that permits identification of the Enterobacteriaceae and other gram negative, oxidase negative bacteria. The system allows the simultaneous inoculation of all media present in the sectors and the execution of 15 biochemical reactions. Microorganism is identified evaluating the colour change of the different culture media after 18 to 24 hours of incubation at 36 ±1 °C and by a code number obtained from biochemical reaction interpretation. Oxidase, Kovacs Reagent Indol (e.g. 1.00181.0002, 1.09293.0100, 1.11350.0001) and Codebook are not included.
  • Based on biochemical tests performed on culture media containing specific substrates
  • Combination of positive and negative permits identification of bacteria by using the Codebook
Product Family Options
Product Information
  • Beskrivelse
    Pakn
    Availability
    Price
  • Enterosystem 18 R
    20 Tests
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.090,00 kr
    1.090,00 kr
    1 * 1 Each
       1.090,00 kr
  • Listeria system 18R
    20 Tests
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.090,00 kr
    1.090,00 kr
    1 * 1 Each
       1.090,00 kr
  • Bacterial Code Book for Enterobacteriaceae identification using Oxi/Ferm Pluri Test
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 312,00 kr
    312,00 kr
    1 * 1 Items
       312,00 kr
  • EnteroPluri-Test Codebook, bacterial code book for Enterobacteriaceae identification
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 324,00 kr
    324,00 kr
    1 * 1 Items
       324,00 kr
  • Strepto system 12R
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.220,00 kr
    1.220,00 kr
    1 * 1 Items
       1.220,00 kr
  • EnteroPluri-Test, system with 12 agar media by simultaneous inoculum for Enterobacteriaceae identification
    10 Tests
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 823,00 kr
    823,00 kr
    1 * 1 Each
       823,00 kr
  • EnteroPluri-Test, system with 12 agar media by simultaneous inoculum for Enterobacteriaceae identification
    25 Tests
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.890,00 kr
    1.890,00 kr
    1 * 1 Each
       1.890,00 kr
  • System for the identification of Gram negative, oxidase positive bacteria
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 823,00 kr
    823,00 kr
    1 * 1 Items
       823,00 kr
  • System for the identification of Gram negative, oxidase positive bacteria
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.890,00 kr
    1.890,00 kr
    1 * 1 Items
       1.890,00 kr
  • Additional biochemical reagents for Entero System 18 R
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 604,00 kr
    604,00 kr
    1 * 1 Each
       604,00 kr
  • Additional biochemical reagents for Listeria System 18
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 882,00 kr
    882,00 kr
    1 * 1 Each
       882,00 kr
  • Additional biochemical reagents for Staph identification systems
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.110,00 kr
    1.110,00 kr
    1 * 1 Each
       1.110,00 kr
  • V.P. TEST Reagent and NaOH and alpha-naphthol solutions for Voges Proskauer test
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 799,00 kr
    799,00 kr
    1 * 1 Each
       799,00 kr
  • “Code book” manual for Enterosystem 18R
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 198,00 kr
    198,00 kr
    1 * 1 Items
       198,00 kr
  • Enterosystem 24R: Biochemical identification of enterobacteria
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.360,00 kr
    1.360,00 kr
    1 * 1 Items
       1.360,00 kr
  • Anaerobe system - biochemical identification of anaerobic microorganisms
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 1.770,00 kr
    1.770,00 kr
    1 * 20 Items
       1.770,00 kr
  • Enterosystem 18R: Biochemical identification of enterobacteria
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 258,00 kr
    258,00 kr
    1 * 1 Each
       258,00 kr