VWR® Coagulase Test
Supplier: VWR Chemicals
About this item
Confirmative test for coagulase-positive Staphylococcus.
- Formulated according to ISO 6888-1
- Can be used to differentiate Staphylococcus aureus from coagulase-negative Staphylococci
2 Options Available Below
Product Details & Documents
Confirmative test for coagulase-positive Staphylococcus.
- Formulated according to ISO 6888-1
- Can be used to differentiate Staphylococcus aureus from coagulase-negative Staphylococci
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 673030ZL
- 673333ZL
- Shelf life:
- 720
- 1080 days (Ampoule)
- Opbevaringstemperatur:
- 2...8 °C
- 2...8 °C
Specifications
Product Family Options
Product Information
- BeskrivelsePaknAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.673030ZLShelf life720Opbevaringstemperatur2...8 °C - Specifications:
More
Specifications
Cat. No.673333ZLShelf life1080 days (Ampoule)Opbevaringstemperatur2...8 °C