Orders LinkContactUsLinkComponent
Dial thermometer for swimming pool
Dial thermometer for swimming pool
Dial thermometer for swimming pool, red filling, 0...+40 °C, 190 mm
Katalog # AMARZ640870
Leverandør:   Amarell
Dial thermometer for swimming pool
Katalog # AMARZ640870
Leverandør:  Amarell
Dial thermometer for swimming pool
Dial thermometer for swimming pool, red filling, 0...+40 °C, 190 mm
Price
887,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Special thermometer used for measuring the temperatures from the swimming pool.
Specifikationer
  • Ø:
    190 mm
  • Beskrivelse:
    Dial thermometer for swimming pool, red filling
  • Område:
    0...+40 °C