Orders LinkContactUsLinkComponent
Process documentaion
Leverandør:   MIELE
Process documentaion
Leverandør:  MIELE
Certifikater
Process documentaion
2 tilgængelige muligheder nedenfor
Product Details & Documents
Specifications for Product Family
Specifikationer
  • Catalog No:
  • MIEL10019160
  • MIEL10019220
  • 141-0631
  • MIEL10106600
  • 141-0632
  • B×D×H:
  • Ø 6×130 mm
  • Ø 2.5×80 mm
Specifikationer
Product Family Options
Produktinformation
  • Til
    Tilgængelighed
    Price
  • MIEL10019160
    APS 101 Sego, Segosoft Miele Edition (additional licences) for connection with a second or additional appliance, Ø 6×130 mm
    APS 101 Sego, Segosoft Miele Edition (additional licences) for connection with a second or additional appliance, Ø 6×130 mm
    All glassware washers
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 5.720,00 kr
    5.720,00 kr
    1 * 1 Items
       5.720,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MIEL10019160
    B×D×H
    Ø 6×130 mm
  • MIEL10019220
    Segosoft Miele edition, solution USB: process documentation software for data transfer using USB stick: Comfort software, installation instructions, USB data logger module with 1,4 m lead and interface seriel cable to connect the unit and the recorder (3 m), USB device
    Connection: 1 unit
    W×D×H: 109×90×31 mm
    Segosoft Miele edition, solution USB: process documentation software for data transfer using USB stick: Comfort software, installation instructions, USB data logger module with 1,4 m lead and interface seriel cable to connect the unit and the recorder (3 m), USB device
    Connection: 1 unit
    W×D×H: 109×90×31 mm
    G 788 (inc CD), PG 8583, PG 8583 CD, PG 8593
    Discontinued
    Specifikationer:
    More
    Specifications
    Cat. No.
    MIEL10019220
    B×D×H
  • 141-0631
    Segosoft Miele edition, USB update (Upgrade Comfort Plus)
    Segosoft Miele edition, USB update (Upgrade Comfort Plus)
    All glassware washers
    Discontinued
    Specifikationer:
    More
    Specifications
    Cat. No.
    141-0631
    B×D×H
  • MIEL10106600
    APS 102 Sego, SegoAssign Plus To connect the Segosoft with the in-house patient management system, Ø 2,5×80 mm
    APS 102 Sego, SegoAssign Plus To connect the Segosoft with the in-house patient management system, Ø 2,5×80 mm
    All glassware washers
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 6.690,00 kr
    6.690,00 kr
    1 * 1 Items
       6.690,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MIEL10106600
    B×D×H
    Ø 2.5×80 mm
  • 141-0632
    APH 330 adapter Gender Changer for connection of two homopolar electrical connections of HAWO sealing devices
    APH 330 adapter Gender Changer for connection of two homopolar electrical connections of HAWO sealing devices
    G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
    Discontinued
    Specifikationer:
    More
    Specifications
    Cat. No.
    141-0632
    B×D×H