Anti-RAF Mouse Monoclonal Antibody [clone: 3E2]
Catalog # USBIR0495-05B
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen symbol:RAF
- Clonality:Monoclonal
- Clone:3E2
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
Type: Primary
Antigen: RAF
Clonality: monoclonal
Clone: 300
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human