Order Entry
Denmark
ContactUsLinkComponent
 
Anti-Parvovirus B19, Human Mouse Monoclonal Antibody [clone: 2Q2041]
  USBIP3113-81C
Anti-Parvovirus B19, Human Mouse Monoclonal Antibody [clone: 2Q2041]
  USBIP3113-81C

 

  • Antistof type:
    Primary
  • Antigen name:
    Parvovirus B19, Human
  • Clonality:
    Monoclonal
  • Clone:
    2Q2041
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Vært:
    Mouse
  • ImmunFluorescens:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,2MPBS, pH 7.6, 0,1% sodium azide.
  • Opbevaringstemperatur:
    Cool
  • Koncentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Størrelse:
    100 µg
  • Pakn:
    100 µG

 

 

Recommended dilution: ELISA: 1:200; Immunofluorescence: 1:200-1:400; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: Parvovirus B19, Human
Clonality: Monoclonal
Clone: 2Q2041
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human