Anti-Myt1, Myelin transcription factor 1, Proteolipid protein binding protein Rabbit Polyclonal Antibody
USBIM9800-80C
- Antistof type:Primary
- Antigen name:Myelin transcription factor 1
- Antigen symbol:MYT1
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:liquid
- Antigen synonymer:Proteolipid Protein binding Protein
- Storage buffer:PBS, 0,01% BSA, 0,5% azide, 0,001% thimerosal.
- Opbevaringstemperatur:Cool
- Koncentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Størrelse:100 µg
- Pakn:100 µG
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Myt1, Myelin transcription factor 1, Proteolipid protein binding protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat