Anti-IgG Donkey Polyclonal Antibody (Gold, 12nm)
Katalog # USBII1903-80
Leverandør: US Biological
Specifikationer
- Antistof type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Konjugation:Gold, 12nm
- ELISA:Yes
- Vært:Donkey
- Isotype:IgG
- Reactivity:Chicken
- Western blot:Yes
- Form:liquid
- Storage buffer:0,01M sodium borate/sodium phosphate, 0,15Msodium chloride, pH 8.5, 15 mg/ml BSA (IgG-Free, Protease-Free), 0,05% sodium azide.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Tested applications:Immunoelectrophoresis, Electron Microscopy
- Purification:purified by immunoaffinity chromatography
- Størrelse:300 µl
- Pakn:300 µl
Specifikationer
Om denne vare
Recommended dilution: ELISA: 1:20-1:40; Immunoelectrophoresis: 1:20-1:40; Electron MImmunochemistryroscopy: 1:20-1:40
Do not freeze!
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: 12 nm Gold
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Chicken