Anti-Humanin 1 Goat Polyclonal Antibody
USBIH7953-30Q
- Antistof type:Primary
- Antigen name:Humanin 1
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Goat
- Reactivity:Human
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sekvens:C-TSEIDLPVKRRT
- Opbevaringstemperatur:Cool
- Koncentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Størrelse:100 µg
- Pakn:100 µG
Recommended dilution: ELISA: 1:16000
Type: Primary
Antigen: Humanin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human