Anti-HIV-2 gag p26 Mouse Monoclonal Antibody [clone: 2C10/E10]
Katalog # USBIH6009-21
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen symbol:HIV-2 gag p26
- Clonality:Monoclonal
- Clone:2C10/E10
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:0,1M Tris-HCl, pH 7,5.
- Opbevaringstemperatur:Cool
- Koncentration:~0,25 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Størrelse:100 µg
- Pakn:100 µG
Specifikationer
Om denne vare
Type: Primary
Antigen: HIV-2 gag p26
Clonality: monoclonal
Clone: 2C10/E10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: