Anti-Heat Shock Protein 47 Mouse Monoclonal Antibody [clone: 4E122]
Katalog # USBIH1834-20
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:Heat Shock Protein 47
- Antigen symbol:HSP47
- Clonality:Monoclonal
- Clone:4E122
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Mouse
- Isotype:IgG2b
- Reactivity:Hamster,Mouse,Pig,Rabbit,Monkey,Sheep,Chicken,Human,Bovine,Guinea pig,Dog,Rat
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide, 50% glycerol.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Størrelse:50 µg
- Pakn:100 µG
Specifikationer
Om denne vare
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50
Type: Primary
Antigen: Heat Shock Protein 47
Clonality: monoclonal
Clone: 4E+122
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Chicken, Dog, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep