Anti-FZD7 Rabbit polyclonal antibody
Catalog # USBIF6401-30C
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Frizzled 7
- Antigen symbol:FZD7
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitop:C-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Størrelse:50 µg
- Pakn:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 15 ?g/ml.
Type: Primary
Antigen: FZD7 (frizzled class receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human