Anti-Fibrinogen Goat Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIF4199-92J
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Fibrinogen
- Clonality:Polyclonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Vært:Goat
- Isotype:IgG
- Reactivity:Rat
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by DEAE chromatography
- Størrelse:1 ml
- Pakn:1 mL
Specifications
About this item
Recommended dilution: FLISA: 1:10-1:40; Immunofluorescence: 1:10-1:40
Type: Primary
Antigen: Fibrinogen (fibrinogen)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat