Anti-Estradiol 17b Rabbit Polyclonal Antibody
USBIE3550-11
- Antistof type:Primary
- Antigen name:Estradiol 17b
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Udformning:Neat Serum
- Form:liquid
- Storage buffer:PBS, 40% glycerol
- Opbevaringstemperatur:Cool
- Koncentration:~200,000 tests/ml (RIA DAB system)
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Størrelse:1000 Tests
- Pakn:1.000 Tests
Type: Primary
Antigen: Estradiol 17b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: