Anti-CD53 Mouse Monoclonal Antibody [clone: 3H1540]
USBIC2408-11
- Antistof type:Primary
- Antigen name:CD53 molecule
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:3H1540
- Konjugation:Unconjugated
- Flowcytometri:Yes
- Vært:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Rat
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Størrelse:25 mg
- Pakn:250 µG
Type: Primary
Antigen: CD53 (CD53 molecule)
Clonality: Monoclonal
Clone: 3H1540
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat