Anti-ITGAX Hamster Monoclonal Antibody [clone: N418]
Katalog # USBIC2262-57D1
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:integrin, alpha X (complement component 3 receptor 4 subunit)
- Antigen symbol:ITGAX
- Clonality:Monoclonal
- Clone:N418
- Konjugation:Unconjugated
- Flowcytometri:Yes
- Vært:Hamster
- ImmunFluorescens:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Størrelse:250 µg
- Pakn:250 µG
Specifikationer
Om denne vare
Recommended dilution: Flow Cytometry: 1:25-1:100
Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: Monoclonal
Clone: N418
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse