Anti-ACE/AGT Rabbit Polyclonal Antibody
Katalog # USBIA2294-48A5
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:Angiotensin I, II
- Antigen symbol:ACE/AGT
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Form:lyophilized powder
- Antigen synonymer:ACE/AGT
- Aminosyretal:1 to 7
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified
- Størrelse:200 µg
- Pakn:200 µG
Specifikationer
Om denne vare
Type: Primary
Antigen: ACE/AGT (Angiotensin I, II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat