Anti-ANGPT2 Rabbit Polyclonal Antibody
Catalog # USBIA2293-10
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Angiopoietin 2
- Antigen symbol:ANGPT2
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Udformning:Neat Serum
- Form:lyophilized powder
- Antigen synonymer:ANG2
- Storage buffer:0,05% sodium azide.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Størrelse:100 µl
- Pakn:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:10000-1:100000; Western Blot: 1:1000-1:5000
Type: Primary
Antigen: ANGPT2 (angiopoietin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse