Anti-ADRB2 Chicken Polyclonal Antibody
Catalog # USBIA0906-88B
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Adrenoceptor beta 2, surface
- Antigen symbol:ADRB2
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Chicken
- Isotype:IgY
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Aminosyretal:335 to 408
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Størrelse:50 µg
- Pakn:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:200
Type: Primary
Antigen: ADRB2 (adrenoceptor beta 2, surface)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human