Anti-STMN1 Rabbit Polyclonal Antibody
USBI146168
- Antistof type:Primary
- Antigen name:Stathmin 1
- Antigen symbol:STMN1
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Rabbit
- ImmunFluorescens:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitop:C-terminal
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Størrelse:100 µg
- Pakn:100 µG
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: STMN1 (stathmin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse