Anti-ICAM1 Mouse Monoclonal Antibody [clone: 13k24]
USBI143151
- Antistof type:Primary
- Antigen name:intercellular adhesion molecule 1
- Antigen symbol:ICAM1
- Clonality:Monoclonal
- Clone:13k24
- Konjugation:Unconjugated
- Vært:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide, 50% glycerol.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Størrelse:100 µl
- Pakn:100 µl
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:200
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone: 13k24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human