Order Entry
Denmark
ContactUsLinkComponent
 
Anti-DNASE1 Mouse Monoclonal Antibody [clone: 13j29]
  USBI140050
Anti-DNASE1 Mouse Monoclonal Antibody [clone: 13j29]
  USBI140050

 

  • Antistof type:
    Primary
  • Antigen name:
    deoxyribonuclease I
  • Antigen symbol:
    DNASE1
  • Clonality:
    Monoclonal
  • Clone:
    13j29
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Vært:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
  • Opbevaringstemperatur:
    Cool
  • Koncentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Størrelse:
    200 µg
  • Pakn:
    200 µG

 

 

Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: DNASE1 (deoxyribonuclease I)
Clonality: monoclonal
Clone: 13j29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human