Anti-NEK10 Mouse Monoclonal Antibody [clone: 1C9]
Catalog # USBI130269
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:NIMA-related kinase 10
- Antigen symbol:NEK10
- Clonality:Monoclonal
- Clone:1C9
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Mouse
- ImmunFluorescens:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Aminosyretal:211 to 301
- Storage buffer:PBS, pH 7,2.
- Sekvens:RYFMEANRNTVTCHHELAVLSHETFEKASLSSSSSGAASLKSELSESADLPPEGFQASYGKDEDRACDEILSDDNFNLENAEKDTYSEVD
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:100 µg
- Pakn:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: NEK10 (NIMA-related kinase 10)
Clonality: monoclonal
Clone: 1C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse