Anti-TNFRSF9 Rat Monoclonal Antibody [clone: M4202]
Katalog # USBI044645
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 9
- Antigen symbol:TNFRSF9
- Clonality:Monoclonal
- Clone:M4202
- Konjugation:Unconjugated
- Flowcytometri:Yes
- Vært:Rat
- Isotype:IgG1 kappa
- Form:liquid
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Opbevaringstemperatur:Cool
- Koncentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:50 µg
- Pakn:50 µG
Specifikationer
Om denne vare
Type: Primary
Antigen: TNFRSF9 (tumor necrosis factor receptor superfamily, member 9)
Clonality: Monoclonal
Clone: M4202
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: