Anti-TCEA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI042681-HRP
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Transcription Elongation Factor A (SII), 1
- Antigen symbol:TCEA1
- Clonality:Polyclonal
- Konjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitop:internal
- Form:liquid
- Aminosyretal:97 to 126
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: TCEA1 (transcription elongation factor A (SII), 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human