Anti-STK25 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI042401-PE
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Serine/threonine Kinase 25
- Antigen symbol:STK25
- Clonality:Polyclonal
- Konjugation:PE (Phycoerythrin)
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitop:C-terminal
- Form:liquid
- Aminosyretal:326 to 355
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: STK25 (serine/threonine kinase 25)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human