Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Anti-SRC8 Rabbit Polyclonal Antibody

Anti-SRC8 Rabbit Polyclonal Antibody

200 µl
Katalog # USBI042293
Leverandør:   US Biological
Anti-SRC8 Rabbit Polyclonal Antibody
Katalog # USBI042293
Leverandør:  US Biological
Anti-SRC8 Rabbit Polyclonal Antibody
Anti-SRC8 Rabbit Polyclonal Antibody

200 µl

Price
5.140,00 kr
Log ind for tilgængelighed
1 * 200 µl
Product Details & Documents

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: SRC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Specifikationer
  • Størrelse:
    200 µl
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    purified by Protein A chromatography
  • Konjugation:
    Unconjugated
  • Form:
    liquid
  • Isotype:
    IgG
  • Aminosyretal:
    16 to 46
  • Epitop:
    N-terminal
  • Shipping temperature:
    Ice
  • Cat. No.:
    USBI042293
  • Reactivity:
    Human
  • Vært:
    Rabbit
  • Antistof type:
    Primary
  • Opbevaringstemperatur:
    Cool
  • Antigen symbol:
    SRC8
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • ELISA:
    Yes