Anti-POMK Rabbit Polyclonal Antibody (Biotin)
Katalog # USBI041657-BIOTIN
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:protein-O-mannose kinase
- Antigen symbol:POMK
- Clonality:Polyclonal
- Konjugation:Biotin
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitop:C-terminal
- Form:liquid
- Aminosyretal:244 to 273
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifikationer
Om denne vare
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: POMK (protein-O-mannose kinase)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse