Anti-SBK3 Rabbit Polyclonal Antibody (Biotin)
USBI041656-BIOTIN
- Antistof type:Primary
- Antigen name:SH3 domain binding kinase family, member 3
- Antigen symbol:SBK3
- Clonality:Polyclonal
- Konjugation:Biotin
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitop:C-terminal
- Form:liquid
- Aminosyretal:331 to 360
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SBK3 (SH3 domain binding kinase family, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse