Order Entry
Denmark
ContactUsLinkComponent
Anti-SGCD Rabbit Polyclonal Antibody
Anti-SGCD Rabbit Polyclonal Antibody
  USBI041648
Anti-SGCD Rabbit Polyclonal Antibody
  USBI041648

 

  • Antistof type:
    Primary
  • Antigen name:
    Sarcoglycan, delta
  • Antigen symbol:
    SGCD
  • Clonality:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Vært:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Form:
    liquid
  • Aminosyretal:
    231 to 261
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SGCD (sarcoglycan, delta (35kDa dystrophin-associated glycoprotein))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse