Anti-RPL8 Rabbit Polyclonal Antibody (Biotin)
USBI041218-BIOTIN
- Antistof type:Primary
- Antigen name:Ribosomal Protein L8
- Antigen symbol:RPL8
- Clonality:Polyclonal
- Konjugation:Biotin
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitop:C-terminal
- Form:liquid
- Aminosyretal:206 to 236
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: RPL8 (ribosomal protein L8)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human