Anti-PTPN1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # USBI040658-HRP
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:Protein tyrosine phosphatase, non-receptor type 1
- Antigen symbol:PTPN1
- Clonality:Polyclonal
- Konjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitop:internal
- Form:liquid
- Aminosyretal:221 to 249
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifikationer
Om denne vare
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: PTPN1 (protein tyrosine phosphatase, non-receptor type 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse