Anti-MAP1ALC3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
USBI038090-HRP
- Antistof type:Primary
- Antigen symbol:MAP1ALC3
- Clonality:Polyclonal
- Konjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100
Type: Primary
Antigen: MAP1ALC3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse