Anti-KRT6A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalog # USBI037623-FITC
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:keratin 6A
- Antigen symbol:KRT6A
- Clonality:Polyclonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitop:internal
- Form:liquid
- Aminosyretal:212 to 240
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifikationer
Om denne vare
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: KRT6A (keratin 6A)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse