Order Entry
Denmark
ContactUsLinkComponent
Anti-FIGNL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-FIGNL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI035658-FITC
Supplier:  US Biological
undefined
Anti-FIGNL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI035658-FITC
Supplier:  US Biological

Specifications

  • Antistof type:
    Primary
  • Antigen name:
    Fidgetin-like 1
  • Antigen symbol:
    FIGNL1
  • Clonality:
    Polyclonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Vært:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Form:
    liquid
  • Aminosyretal:
    11 to 40
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: FIGNL1 (fidgetin-like 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human