Order Entry
Denmark
ContactUsLinkComponent
Anti-FAM83A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-FAM83A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Katalog # USBI035490-APC
Leverandør:  US Biological
undefined
Anti-FAM83A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Katalog # USBI035490-APC
Leverandør:  US Biological

Specifikationer

  • Antistof type:
    Primary
  • Antigen name:
    Family with sequence similarity 83, member A
  • Antigen symbol:
    FAM83A
  • Clonality:
    Polyclonal
  • Konjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Vært:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitop:
    internal
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

Specifikationer

Om denne vare

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: FAM83A (family with sequence similarity 83, member A)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human