Order Entry
Denmark
ContactUsLinkComponent
Anti-FAM69B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-FAM69B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI035475-FITC
Supplier:  US Biological
undefined
Anti-FAM69B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI035475-FITC
Supplier:  US Biological

Specifications

  • Antistof type:
    Primary
  • Antigen name:
    Family with sequence similarity 69, member B
  • Antigen symbol:
    FAM69B
  • Clonality:
    Polyclonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Vært:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Form:
    liquid
  • Aminosyretal:
    114 to 144
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: FAM69B (family with sequence similarity 69, member B)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse